Oxcarbazepine ≥98%

Supplier: Ambeed
Warning

Synonyms: 10-Oxo-10,11-dihydro-5H-dibenzo[b,f]azepine-5-carboxamide, 10,11-Dihydro-10-oxo-5H-dibenzo[b,f]azepine-5-carboxamide

A127391-25G A127391-5G A127391-100G A127391-1G A127391-250MG A127391-50MG
77592-476EA 59.45 USD
77592-476 77592-478 77685-106 77685-107 77685-108 77685-109
Oxcarbazepine ≥98%
Oxcarbazepine

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂O₂
MW: 252.27 g/mol
MDL Number: MFCD00865307
CAS Number: 28721-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR