Dibromo(3,4,7,8-tetramethyl-1,10-phenanthroline-κN1,κN10)-Nickel ≥98%

Supplier: Ambeed
Danger

A1272895-100MG A1272895-250MG
77596-648EA 22.55 USD
77596-648 77596-650
Dibromo(3,4,7,8-tetramethyl-1,10-phenanthroline-κN1,κN10)-Nickel ≥98%
Dibromo(3,4,7,8-tetramethyl-1,10-phenanthroline-κN1,κN10)-Nickel

Precaución: for Research Use Only

Formula: C₁₆H₁₆Br₂N₂Ni
MW: 454.8130 g/mol
CAS Number: 2414273-20-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR