(T-4)-Dichloro(2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-N1,N10)-Nickel ≥98%

Supplier: Ambeed
Danger

A1272893-250MG A1272893-1G
77596-646EA 52.28 USD
77596-646 77621-424
(T-4)-Dichloro(2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-N1,N10)-Nickel ≥98%
(T-4)-Dichloro(2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-N1,N10)-Nickel

Precaución: for Research Use Only

Formula: C₂₆H₂₀Cl₂N₂Ni
MW: 490.0498 g/mol
CAS Number: 120763-13-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR