1,2-Bis(4-iodophenyl)diazene ≥98%

Supplier: Ambeed
Warning

A1272862-250MG A1272862-1G
77621-420EA 78.28 USD
77621-420 77867-143
1,2-Bis(4-iodophenyl)diazene ≥98%
1,2-Bis(4-iodophenyl)diazene

Precaución: for Research Use Only

Formula: C₁₂H₈I₂N₂
MDL Number: MFCD00185033
CAS Number: 1601-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR