2,9-Dibromo-1,10-phenanthroline-5,6-dione ≥97%

Supplier: Ambeed

A1271729-100MG A1271729-250MG
77867-138EA 37 USD
77867-138 77867-139
2,9-Dibromo-1,10-phenanthroline-5,6-dione ≥97%
2,9-Dibromo-1,10-phenanthroline-5,6-dione

Precaución: for Research Use Only

Formula: C₁₂H₄Br₂N₂O₂
MDL Number: MFCD32709438
CAS Number: 943861-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR