3-Methoxy-5-nitrobenzene-1,2-diamine ≥97%

Supplier: Ambeed
Warning

A1271197-100MG A1271197-50MG
77592-466EA 243.95 USD
77592-466 77621-404
3-Methoxy-5-nitrobenzene-1,2-diamine ≥97%
3-Methoxy-5-nitrobenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₇H₉N₃O₃
MDL Number: MFCD25965730
CAS Number: 64381-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR