5-Chloro-2-methylbenzothiazole ≥99%

Supplier: Ambeed
Warning

A126818-5G A126818-25G
77571-592EA 32.96 USD
77571-592 77670-788
5-Chloro-2-methylbenzothiazole ≥99%
5-Chloro-2-methylbenzothiazole

Precaución: for Research Use Only

Formula: C₈H₆ClNS
MW: 183.66 g/mol
MDL Number: MFCD00022881
CAS Number: 1006-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR