6-Methoxyflavone ≥97%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-2-phenyl-4H-1-benzopyran-4-one, 6-Methoxy-2-phenyl-4H-benzo[b]pyran-4-one, 6-Methoxy-2-phenyl-4-benzopyrone, 6-Methoxy-2-phenyl-4H-chromen-4-one, 6-Methoxy-2-phenylchromen-4-one

A126806-5G A126806-1G
77599-840EA 93.28 USD
77599-840 77624-060
6-Methoxyflavone ≥97%
6-Methoxyflavone

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₃
MW: 252.27 g/mol
MDL Number: MFCD00017322
CAS Number: 26964-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR