Bismuth tripotassium dicitrate Bi 35.0-38.5%

Supplier: Ambeed
Warning

Synonyms: Bismuth(III) tripotassium 2-hydroxypropane-1,2,3-tricarboxylate, Bismuth potassium citrate

A126800-10G A126800-1G A126800-5G
77876-911EA 13 USD
77876-911 77876-912 77876-913
Bismuth tripotassium dicitrate Bi 35.0-38.5%
Bismuth tripotassium dicitrate

Precaución: for Research Use Only

Formula: C₁₂H₁₀BiK₃O₁₄
MDL Number: MFCD02101411
CAS Number: 57644-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR