(1-(tert-Butoxycarbonyl)-6-((tert-butyldimethylsilyl)oxy)-1H-indol-2-yl)boronic acid ≥98%

Supplier: Ambeed

A1264464-100MG A1264464-1G A1264464-250MG A1264464-5G
77867-116EA 111 USD
77867-116 77867-117 77867-118 77867-119
(1-(tert-Butoxycarbonyl)-6-((tert-butyldimethylsilyl)oxy)-1H-indol-2-yl)boronic acid ≥98%
(1-(tert-Butoxycarbonyl)-6-((tert-butyldimethylsilyl)oxy)-1H-indol-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₉H₃₀BNO₅Si
MW: 391.34 g/mol
MDL Number: MFCD11616318
CAS Number: 848357-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR