3-(5-Fluorobenzofuran-3-yl)-4-(5-methyl-5H-[1,3]dioxolo[4,5-f]indol-7-yl)-1H-pyrrole-2,5-dione ≥97%

Supplier: Ambeed
Warning

Synonyms: 9-ING-41

A1264331-25MG A1264331-100MG
77571-576EA 85.49 USD
77571-576 77621-382
3-(5-Fluorobenzofuran-3-yl)-4-(5-methyl-5H-[1,3]dioxolo[4,5-f]indol-7-yl)-1H-pyrrole-2,5-dione ≥97%
3-(5-Fluorobenzofuran-3-yl)-4-(5-methyl-5H-[1,3]dioxolo[4,5-f]indol-7-yl)-1H-pyrrole-2,5-dione

Precaución: for Research Use Only

Formula: C₂₂H₁₃FN₂O₅
MW: 404.35 g/mol
MDL Number: MFCD16495778
CAS Number: 1034895-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR