Isopropyl 2-((5-acrylamido-4-((2-(dimethylamino)ethyl)(methyl)amino)-2-methoxyphenyl)amino)-4-(1-methyl-1H-indol-3-yl)pyrimidine-5-carboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: Mobocertinib

A1263841-1G A1263841-250MG
77621-376EA 431.57 USD
77621-376 77621-378
Isopropyl 2-((5-acrylamido-4-((2-(dimethylamino)ethyl)(methyl)amino)-2-methoxyphenyl)amino)-4-(1-methyl-1H-indol-3-yl)pyrimidine-5-carboxylate ≥98%
Isopropyl 2-((5-acrylamido-4-((2-(dimethylamino)ethyl)(methyl)amino)-2-methoxyphenyl)amino)-4-(1-methyl-1H-indol-3-yl)pyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₃₂H₃₉N₇O₄
MW: 585.7 g/mol
MDL Number: MFCD32669806
CAS Number: 1847461-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR