(4S,4'S)-4,4'-Di(heptan-4-yl)-4,4',5,5'-tetrahydro-2,2'-bioxazole ≥97%

Supplier: Ambeed
Warning

A1263809-250MG A1263809-1G A1263809-100MG
77571-572EA 350 USD
77571-572 77596-634 77621-374
(4S,4'S)-4,4'-Di(heptan-4-yl)-4,4',5,5'-tetrahydro-2,2'-bioxazole ≥97%
(4S,4'S)-4,4'-Di(heptan-4-yl)-4,4',5,5'-tetrahydro-2,2'-bioxazole

Precaución: for Research Use Only

Formula: C₂₀H₃₆N₂O₂
CAS Number: 2094476-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR