1,6-Dimethylpyrene ≥97%

Supplier: Ambeed
Warning

A1263312-100MG A1263312-1G
77867-099EA 119 USD
77867-099 77867-100
1,6-Dimethylpyrene ≥97%
1,6-Dimethylpyrene

Precaución: for Research Use Only

Formula: C₁₈H₁₄
CAS Number: 27984-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR