[Ru(p-cymene)(MeCN)3](SbF6)2 ≥97%

Supplier: Ambeed
Danger

A1258675-100MG A1258675-250MG A1258675-1G
77596-628EA 100.45 USD
77596-628 77596-630 77621-360
[Ru(p-cymene)(MeCN)3](SbF6)2 ≥97%
[Ru(p-cymene)(MeCN)3](SbF6)2

Precaución: for Research Use Only

Formula: C₁₆H₂₃F₁₂N₃RuSb₂
MW: 829.95 g/mol
CAS Number: 1867114-28-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR