4H,6H-Indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium, 5a,10b-dihydro-2-[2,4,6-tris(1-methylethyl)phenyl]-, (5aS,10bR)-, tetrafluoroborate(1-) (1:1) ≥97%

Supplier: Ambeed
Danger

Synonyms: (5aS,10bR)-2-(2,4,6-Triisopropylphenyl)-5a,10b-dihydro-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate

A1258459-250MG A1258459-100MG A1258459-1G A1258459-50MG
77596-620EA 168.1 USD
77596-620 77621-350 77867-080 77867-081
4H,6H-Indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium, 5a,10b-dihydro-2-[2,4,6-tris(1-methylethyl)phenyl]-, (5aS,10bR)-, tetrafluoroborate(1-) (1:1) ≥97%
4H,6H-Indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium, 5a,10b-dihydro-2-[2,4,6-tris(1-methylethyl)phenyl]-, (5aS,10bR)-, tetrafluoroborate(1-) (1:1)

Precaución: for Research Use Only

Formula: C₂₇H₃₄BF₄N₃O
CAS Number: 1827720-97-7
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR