4-(5-Phenyl-3-(3-(3-(4-(trifluoromethyl)phenyl)ureido)propyl)-1H-pyrazol-1-yl)benzenesulfonamide ≥97%

Supplier: Ambeed
Warning

Synonyms: PTUPB

A1258087-100MG A1258087-25MG A1258087-50MG
77621-328EA 222.48 USD
77621-328 77621-330 77621-332
4-(5-Phenyl-3-(3-(3-(4-(trifluoromethyl)phenyl)ureido)propyl)-1H-pyrazol-1-yl)benzenesulfonamide ≥97%
4-(5-Phenyl-3-(3-(3-(4-(trifluoromethyl)phenyl)ureido)propyl)-1H-pyrazol-1-yl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₂₆H₂₄F₃N₅O₃S
MW: 543.56 g/mol
MDL Number: MFCD32701914
CAS Number: 1287761-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR