(E)-4-((3-Chlorophenyl)diazenyl)-5-hydroxy-3-(4-nitrophenyl)-1H-pyrazole-1-carbothioamide ≥97%

Supplier: Ambeed

Synonyms: Hck-IN-1

A1258085-10MG A1258085-1MG A1258085-5MG
77596-612EA 69.7 USD
77596-612 77596-614 77596-616
(E)-4-((3-Chlorophenyl)diazenyl)-5-hydroxy-3-(4-nitrophenyl)-1H-pyrazole-1-carbothioamide ≥97%
(E)-4-((3-Chlorophenyl)diazenyl)-5-hydroxy-3-(4-nitrophenyl)-1H-pyrazole-1-carbothioamide

Precaución: for Research Use Only

Formula: C₁₆H₁₁ClN₆O₃S
MW: 402.81 g/mol
MDL Number: MFCD32671892
CAS Number: 1473404-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR