1-(1-(Piperidin-4-yl)-1H-pyrazol-4-yl)anthracene-9,10-dione hydrochloride ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
PDK4-IN-1 HCl
A1258083-1MG
A1258083-10MG
A1258083-5MG
77596-610EA
30.75
USD
77596-610
77621-324
77621-326
1-(1-(Piperidin-4-yl)-1H-pyrazol-4-yl)anthracene-9,10-dione hydrochloride ≥97%
1-(1-(Piperidin-4-yl)-1H-pyrazol-4-yl)anthracene-9,10-dione hydrochloride
Precaución: for Research Use Only
Formula:
C₂₂H₂₀ClN₃O₂ MW: 393.8661 g/mol |
CAS Number:
2310262-11-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...