6,6'-Di-tert-butyl-2,2'-bipyridine ≥97%

Supplier: Ambeed
Warning

A1257659-100MG A1257659-250MG A1257659-1G A1257659-5G
77621-318EA 45.32 USD
77621-318 77621-320 77867-067 77867-068
6,6'-Di-tert-butyl-2,2'-bipyridine ≥97%
6,6'-Di-tert-butyl-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₁₈H₂₄N₂
MDL Number: MFCD24449700
CAS Number: 6859-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR