5-Bromo-3-methylisothiazole ≥98%

Supplier: Ambeed
Warning

A125695-250MG A125695-1G A125695-5G
77571-536EA 56 USD
77571-536 77876-906 77876-907
5-Bromo-3-methylisothiazole ≥98%
5-Bromo-3-methylisothiazole

Precaución: for Research Use Only

Formula: C₄H₄BrNS
MDL Number: MFCD01318949
CAS Number: 20493-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR