N-(5-Cyano-2-methylphenyl)acetamide ≥98%

Supplier: Ambeed
Warning

A1255196-1G A1255196-250MG A1255196-5G
77621-282EA 24.72 USD
77621-282 77867-045 77867-046
N-(5-Cyano-2-methylphenyl)acetamide ≥98%
N-(5-Cyano-2-methylphenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MDL Number: MFCD16683000
CAS Number: 81018-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR