4'-Chloro-6-((4-chlorobenzyl)oxy)-3-(1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)-3'-(trifluoromethyl)-[1,1'-biphenyl]-2-ol ≥98%

Supplier: Ambeed
Warning

Synonyms: MYCi975

A1251865-250MG A1251865-50MG
77621-276EA 612.85 USD
77621-276 77621-278
4'-Chloro-6-((4-chlorobenzyl)oxy)-3-(1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)-3'-(trifluoromethyl)-[1,1'-biphenyl]-2-ol ≥98%
4'-Chloro-6-((4-chlorobenzyl)oxy)-3-(1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl)-3'-(trifluoromethyl)-[1,1'-biphenyl]-2-ol

Precaución: for Research Use Only

Formula: C₂₅H₁₆Cl₂F₆N₂O₂
MW: 561.3 g/mol
MDL Number: MFCD32263413
CAS Number: 2289691-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR