Methyl (4-(5,7-difluoro-2-phenyl-1H-indol-3-yl)butanoyl)-L-argininate ≥98%

Supplier: Ambeed
Warning

Synonyms: L-803087

A1251824-1MG A1251824-10MG A1251824-5MG
77596-566EA 42.03 USD
77596-566 77621-264 77621-266
Methyl (4-(5,7-difluoro-2-phenyl-1H-indol-3-yl)butanoyl)-L-argininate ≥98%
Methyl (4-(5,7-difluoro-2-phenyl-1H-indol-3-yl)butanoyl)-L-argininate

Precaución: for Research Use Only

Formula: C₂₅H₂₉F₂N₅O₃
MW: 485.53 g/mol
CAS Number: 217480-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR