2,5-Bis(allyloxy)terephthalic acid ≥97%

Supplier: Ambeed
Warning

A1251661-1G A1251661-5G
77621-260EA 133.9 USD
77621-260 77621-262
2,5-Bis(allyloxy)terephthalic acid ≥97%
2,5-Bis(allyloxy)terephthalic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₆
MW: 278.2574 g/mol
MDL Number: MFCD32856628
CAS Number: 149590-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR