N-((4-(2-(4-Bromo-3-(3-chloro-5-cyanophenoxy)-2-fluorophenyl)acetamido)-3-chlorophenyl)sulfonyl)propionamide ≥97%

Supplier: Ambeed

Synonyms: Elsulphavirine

A1251631-250MG A1251631-25MG
77867-028EA 644 USD
77867-028 77867-029
N-((4-(2-(4-Bromo-3-(3-chloro-5-cyanophenoxy)-2-fluorophenyl)acetamido)-3-chlorophenyl)sulfonyl)propionamide ≥97%
N-((4-(2-(4-Bromo-3-(3-chloro-5-cyanophenoxy)-2-fluorophenyl)acetamido)-3-chlorophenyl)sulfonyl)propionamide

Precaución: for Research Use Only

Formula: C₂₄H₁₇BrCl₂FN₃O₅S
MW: 629.28 g/mol
MDL Number: MFCD32215300
CAS Number: 868046-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR