Fluorescein-DBCO ≥98%

Supplier: Ambeed
Warning

A1251452-1MG A1251452-5MG A1251452-25MG A1251452-50MG
77596-564EA 91.23 USD
77596-564 77621-252 77867-025 77867-026
Fluorescein-DBCO ≥98%
Fluorescein-DBCO

Precaución: for Research Use Only

Formula: C₃₉H₂₇N₃O₆S
MDL Number: MFCD30527400
CAS Number: 2054339-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR