4-((S)-2-(4-Aminobutyl)-35-(4-((4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexanecarboxamido)methyl)-1H-1,2,3-triazol-1-yl)-4,8-dioxo-6,12,15,18,21,24,27,30,33-nonaoxa-3,9-diazapentatriacontanamido)benzyl ((S)-4,11-diethyl-9-hydroxy-3,14-dioxo-3,4,12,14-tetrahydro-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-4-yl) carbonate ≥98%

Supplier: Ambeed
Danger

A1250774-100MG A1250774-10MG
77596-552EA 958.38 USD
77596-552 77621-242
4-((S)-2-(4-Aminobutyl)-35-(4-((4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexanecarboxamido)methyl)-1H-1,2,3-triazol-1-yl)-4,8-dioxo-6,12,15,18,21,24,27,30,33-nonaoxa-3,9-diazapentatriacontanamido)benzyl ((S)-4,11-diethyl-9-hydroxy-3,14-dioxo-3,4,12,14-tetrahydro-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-4-yl) carbonate ≥98%
4-((S)-2-(4-Aminobutyl)-35-(4-((4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexanecarboxamido)methyl)-1H-1,2,3-triazol-1-yl)-4,8-dioxo-6,12,15,18,21,24,27,30,33-nonaoxa-3,9-diazapentatriacontanamido)benzyl ((S)-4,11-diethyl-9-hydroxy-3,14-dioxo-

Precaución: for Research Use Only

Formula: C₇₃H₉₇N₁₁O₂₂
MW: 1480.61 g/mol
MDL Number: MFCD32701926
CAS Number: 1279680-68-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR