(4-Bromobenzyl)boronic acid ≥98%
Supplier: Ambeed
A1249971-100MG
A1249971-250MG
77867-004EA
107
USD
77867-004
77867-005
(4-Bromobenzyl)boronic acid ≥98%
(4-Bromobenzyl)boronic acid
Precaución: for Research Use Only
Formula:
C₇H₈BBrO₂ MW: 214.85 g/mol |
MDL Number:
MFCD18861708 CAS Number: 1350513-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...