4-(1-Cyanocyclopropyl)-2-fluorobenzeneboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

A1249048-250MG A1249048-100MG
77596-524EA 53.3 USD
77596-524 77866-993
4-(1-Cyanocyclopropyl)-2-fluorobenzeneboronic acid pinacol ester ≥98%
4-(1-Cyanocyclopropyl)-2-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₁₉BFNO₂
MDL Number: MFCD22418790
CAS Number: 917397-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR