4-Methylbenzyl isocyanide ≥98%

Supplier: Ambeed
Danger

Synonyms: 1-(Isocyanomethyl)-4-methylbenzene

A1248645-100MG A1248645-250MG
77866-989EA 173 USD
77866-989 77866-990
4-Methylbenzyl isocyanide ≥98%
4-Methylbenzyl isocyanide

Precaución: for Research Use Only

Formula: C₉H₉N
MDL Number: MFCD02664586
CAS Number: 39495-97-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR