(S)-7-Hydroxy-2-(4-hydroxy-2-methoxyphenyl)-5-methoxy-8-((R)-5-methyl-2-(prop-1-en-2-yl)hex-4-en-1-yl)chroman-4-one ≥98%

Supplier: Ambeed
Warning

Synonyms: (2S)-2'-Methoxykurarinone

A124808-1MG A124808-5MG
77685-092EA 141 USD
77685-092 77685-093
(S)-7-Hydroxy-2-(4-hydroxy-2-methoxyphenyl)-5-methoxy-8-((R)-5-methyl-2-(prop-1-en-2-yl)hex-4-en-1-yl)chroman-4-one ≥98%
(S)-7-Hydroxy-2-(4-hydroxy-2-methoxyphenyl)-5-methoxy-8-((R)-5-methyl-2-(prop-1-en-2-yl)hex-4-en-1-yl)chroman-4-one

Precaución: for Research Use Only

Formula: C₂₇H₃₂O₆
MW: 452.54 g/mol
MDL Number: MFCD21333216
CAS Number: 270249-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR