(5-Methyl-6-(N-methylbenzamido)pyridin-3-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A1245995-1G A1245995-25G A1245995-5G
77866-968EA 9 USD
77866-968 77866-969 77866-970
(5-Methyl-6-(N-methylbenzamido)pyridin-3-yl)boronic acid ≥98%
(5-Methyl-6-(N-methylbenzamido)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅BN₂O₃
MDL Number: MFCD32012267
CAS Number: 446299-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR