6-Phenyl-1-hexene ≥98% stabilised

Supplier: Ambeed
Warning


Stabilization: Stabilised with TBC

A1242811-250MG A1242811-1G
77571-460EA 86 USD
77571-460 77866-955
6-Phenyl-1-hexene ≥98% stabilised
6-Phenyl-1-hexene

Precaución: for Research Use Only

Formula: C₁₂H₁₆
MDL Number: MFCD07775147
CAS Number: 1588-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR