(E)-3-(5-Methoxy-2-methyl-1H-indol-3-yl)-1-(pyridin-4-yl)prop-2-en-1-one 98+%

Supplier: Ambeed
Warning

Synonyms: MOMIPP

A1241510-10MG A1241510-1MG A1241510-5MG
77596-516EA 17.43 USD
77596-516 77596-518 77596-520
(E)-3-(5-Methoxy-2-methyl-1H-indol-3-yl)-1-(pyridin-4-yl)prop-2-en-1-one 98+%
(E)-3-(5-Methoxy-2-methyl-1H-indol-3-yl)-1-(pyridin-4-yl)prop-2-en-1-one

Precaución: for Research Use Only

Formula: C₁₈H₁₆N₂O₂
MW: 292.33 g/mol
MDL Number: MFCD34476008
CAS Number: 1363421-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR