N-(9-Ethyl-9H-carbazol-3-yl)-2-((6-oxo-1,6-dihydropyrimidin-2-yl)thio)acetamide ≥98%

Supplier: Ambeed
Warning

Synonyms: PK095

A1234825-10MG A1234825-1MG A1234825-5MG
77596-508EA 469.45 USD
77596-508 77596-510 77596-512
N-(9-Ethyl-9H-carbazol-3-yl)-2-((6-oxo-1,6-dihydropyrimidin-2-yl)thio)acetamide ≥98%
N-(9-Ethyl-9H-carbazol-3-yl)-2-((6-oxo-1,6-dihydropyrimidin-2-yl)thio)acetamide

Precaución: for Research Use Only

Formula: C₂₀H₁₈N₄O₂S
MW: 378.45 g/mol
MDL Number: MFCD09276463
CAS Number: 380314-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR