2,4-Dichloro-5-fluoro-7H-pyrrolo[2,3-d]pyrimidine ≥97%

Supplier: Ambeed
Warning

A123415-100MG A123415-1G
77670-752EA 97 USD
77670-752 77670-754
2,4-Dichloro-5-fluoro-7H-pyrrolo[2,3-d]pyrimidine ≥97%
2,4-Dichloro-5-fluoro-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂FN₃
MW: 206.005 g/mol
MDL Number: MFCD16249867
CAS Number: 1053228-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR