Motesanib ≥98%

Supplier: Ambeed
Warning

Synonyms: N-(3,3-Dimethylindolin-6-yl)-2-((pyridin-4-ylmethyl)amino)nicotinamide, AMG706

A123381-100MG A123381-50MG A123381-1MG
77611-054EA 238.96 USD
77611-054 77611-056 77670-750
Motesanib ≥98%
Motesanib

Precaución: for Research Use Only

Formula: C₂₂H₂₃N₅O
MW: 373.5 g/mol
MDL Number: MFCD10567689
CAS Number: 453562-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR