Chlorobis(cyclooctene)rhodium(I) dimer 98+%

Supplier: Ambeed
Warning

A1233073-100MG A1233073-1G A1233073-250MG
77592-402EA 107.63 USD
77592-402 77621-172 77621-174
Chlorobis(cyclooctene)rhodium(I) dimer 98+%
Chlorobis(cyclooctene)rhodium(I) dimer

Precaución: for Research Use Only

Formula: C₃₂H₅₆Cl₂Rh₂
MDL Number: MFCD00013287
CAS Number: 12279-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR