2-(Pyridazin-4-yl)thiazole-4-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A1232358-100MG A1232358-1G A1232358-250MG
77866-915EA 222 USD
77866-915 77866-916 77866-917
2-(Pyridazin-4-yl)thiazole-4-carboxylic acid ≥98%
2-(Pyridazin-4-yl)thiazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅N₃O₂S
MDL Number: MFCD24498915
CAS Number: 955400-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR