(S)-N-(2-(3-((4-Hydroxy-4-(5-(pyrimidin-2-yl)pyridin-2-yl)cyclohexyl)amino)pyrrolidin-1-yl)-2-oxoethyl)-3-(trifluoromethyl)benzamide ≥98%

Supplier: Ambeed
Warning

Synonyms: (Rac)-PF-4136309

A1231629-10MG A1231629-1MG A1231629-5MG
77596-488EA 107.63 USD
77596-488 77596-490 77596-492
(S)-N-(2-(3-((4-Hydroxy-4-(5-(pyrimidin-2-yl)pyridin-2-yl)cyclohexyl)amino)pyrrolidin-1-yl)-2-oxoethyl)-3-(trifluoromethyl)benzamide ≥98%
(S)-N-(2-(3-((4-Hydroxy-4-(5-(pyrimidin-2-yl)pyridin-2-yl)cyclohexyl)amino)pyrrolidin-1-yl)-2-oxoethyl)-3-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₂₉H₃₁F₃N₆O₃
MDL Number: MFCD22419015
CAS Number: 857679-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR