(2S,3R,4R,5S,6R)-2-(4-Chloro-3-(4-hydroxybenzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol ≥95%

Supplier: Ambeed
Danger

Synonyms: O-Desethyl Dapagliflozin

A1231619-100MG A1231619-250MG
77621-162EA 108.15 USD
77621-162 77621-164
(2S,3R,4R,5S,6R)-2-(4-Chloro-3-(4-hydroxybenzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol ≥95%
(2S,3R,4R,5S,6R)-2-(4-Chloro-3-(4-hydroxybenzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

Precaución: for Research Use Only

Formula: C₁₉H₂₁ClO₆
MW: 380.8195 g/mol
MDL Number: MFCD28387374
CAS Number: 864070-37-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR