2-Amino-5-methylbenzotrifluoride ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-2-(trifluoromethyl)aniline

A123142-1G A123142-250MG
77614-966EA 40.17 USD
77614-966 77745-903
2-Amino-5-methylbenzotrifluoride ≥98%
2-Amino-5-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₈F₃N
MW: 175.1510 g/mol
MDL Number: MFCD01631581
CAS Number: 87617-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR