(6-Fluoro-1H-indol-2-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A1230462-1G A1230462-250MG
77866-900EA 626 USD
77866-900 77866-901
(6-Fluoro-1H-indol-2-yl)boronic acid ≥98%
(6-Fluoro-1H-indol-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₇BFNO₂
MW: 178.96 g/mol
MDL Number: MFCD11858390
CAS Number: 1613450-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR