(S)-3-(1-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid ≥98%

Supplier: Ambeed
Warning

A1230375-250MG A1230375-100MG A1230375-1G
77596-486EA 202.95 USD
77596-486 77866-897 77866-898
(S)-3-(1-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid ≥98%
(S)-3-(1-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MDL Number: MFCD12913673
CAS Number: 1187930-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR