N1-(5-(4-Chlorophenyl)oxazol-2-yl)benzene-1,3-diamine ≥97%

Supplier: Ambeed

A1229053-5MG A1229053-10MG A1229053-1MG
77596-484EA 16.4 USD
77596-484 77596-480 77596-482
N1-(5-(4-Chlorophenyl)oxazol-2-yl)benzene-1,3-diamine ≥97%
N1-(5-(4-Chlorophenyl)oxazol-2-yl)benzene-1,3-diamine

Precaución: for Research Use Only

Formula: C₁₅H₁₂ClN₃O
MDL Number: MFCD32708524
CAS Number: 1807758-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR