tert-Butyl (2R,6R)-2,6-dimethylpiperazine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: (2R,6R)-tert-Butyl 2,6-dimethylpiperazine-1-carboxylate

A122881-1G A122881-100MG A122881-250MG
77571-396EA 534.57 USD
77571-396 77683-102 77683-103
tert-Butyl (2R,6R)-2,6-dimethylpiperazine-1-carboxylate ≥98%
tert-Butyl (2R,6R)-2,6-dimethylpiperazine-1-carboxylate

Precaución: For research use only.

Formula: C₁₁H₂₂N₂O₂
MW: 214.305 g/mol
MDL Number: MFCD11973747
CAS Number: 574007-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR