Sodium 7-(methylthio)-3-nitro-4-oxo-4H-[1,2,4]triazolo[5,1-c][1,2,4]triazin-6-ide dihydrate 99+%

Supplier: Ambeed

A1228484-50MG A1228484-100MG A1228484-250MG A1228484-25MG
77866-880EA 318 USD
77866-880 77866-877 77866-878 77866-879
Sodium 7-(methylthio)-3-nitro-4-oxo-4H-[1,2,4]triazolo[5,1-c][1,2,4]triazin-6-ide dihydrate 99+%
Sodium 7-(methylthio)-3-nitro-4-oxo-4H-[1,2,4]triazolo[5,1-c][1,2,4]triazin-6-ide dihydrate

Precaución: for Research Use Only

Formula: C₅H₇N₆NaO₅S
MDL Number: MFCD30532663
CAS Number: 928659-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR