Kanamycin sulphate >680 u/mg
Supplier: Ambeed
|
Danger
|
Synonyms:
Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate
A122807-100G
A122807-25G
A122807-5G
A122807-10G
A122807-1G
77599-784EA
39.98
USD
77599-784
77599-786
77599-788
77666-764
77878-904
Kanamycin sulphate >680 u/mg
Kanamycin sulphate
Precaución: for Research Use Only
Formula:
C₁₈H₃₈N₄O₁₅S |
MDL Number:
MFCD00070253 CAS Number: 25389-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...