2-Bromo-3,5-dichlorobenzaldehyde ≥98%

Supplier: Ambeed
Warning

A1227412-250MG A1227412-5G
77866-871EA 10 USD
77866-871 77866-872
2-Bromo-3,5-dichlorobenzaldehyde ≥98%
2-Bromo-3,5-dichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O
MDL Number: MFCD31719953
CAS Number: 873983-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR